    Sort by:
  • Search Within Results
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Grade: 97. Melting Point C121-123*. Boiling Point C: NA. C10H6F6O3. 16261-80-6. IRRITANT