SP Wilmad-LabGlass Pour Out Adapter, SP Industries
Supplier: SP Industries
LG-1353-100
14201-578EA 51.15 USD
14201-578
SP Wilmad-LabGlass Pour Out Adapter, SP Industries
Adapter is designed for the easy transfer of liquids.
  • [ST] inner joint
  • Use for easy pouring
  • Made of borosilicate glass
This borosilicate glass pour out adapter has a standard taper 24/40 inner joint to avoid liquid contact with ground zone area of vessels equipped with standard taper outer joints.