ADIPSYN-1059
SYN-1059-M050 SYN-1059-M005 SYN-1059-M001 SYN-1059-M010
102991-716EA 74.77 USD
102991-716 102991-718 102991-722 102991-720
ADIPSYN-1059
PD0325901 is selective and non ATP-competitive MEK inhibitor with an IC(50) of 0.33nM, roughly 500-fold more potent than CI-1040 on the phosphorylation of ERK1 and ERK2.