7-Methoxy-4-methylcoumarin
Supplier: ADIPOGEN CORP MS
CDX-M0046-M100
CDX-M0046-G005
102990-414EA
157.85
USD
102990-414
102990-412
7-Methoxy-4-methylcoumarin
7-Methoxy-4-methylcoumarin
Coumarin derivative. Fluorescent label. Shown to be UVA-phototoxic against F. cultnorum and to have some weak biological activities.
Formula:
C₁₁H₁₀O₃ MW: 190.2 g/mol |
MDL Number:
MFCD00009773 CAS Number: 2555-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...