Isoxathion
Supplier: ADIPOGEN CORP MS
CDX-I0049-M100 CDX-I0049-M500
102990-302EA 841.85 USD
102990-302 102990-300
Isoxathion
Isoxathion
Isoxazole organothiophosphate insecticide. Works as an acetylcholine esterase inhibitor. Compound can be used as analytical reference material.