7-Ethoxy-4-methylcoumarin

Supplier: ADIPOGEN CORP MS

CDX-E0004-G001 CDX-E0004-M100 CDX-E0004-M200
102989-870EA 131.54 USD
102989-870 102989-872 102989-874
7-Ethoxy-4-methylcoumarin
7-Ethoxy-4-methylcoumarin

Fluorescent probe dye.

Formula: C₁₂H₁₂O₃
MDL Number: MFCD00016971
CAS Number: 87-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR