10,10'-Dimethyl-9,9'-biacridinium Dinitrate
Supplier: ADIPOGEN CORP MS
Synonyms:
Lucigenin
CDX-D0068-G010
CDX-D0068-G002
102987-742EA
210.46
USD
102987-742
102987-744
10,10'-Dimethyl-9,9'-biacridinium Dinitrate
Lucigenin
Chemiluminescent probe for the detection of peroxides in biological systems. Reagent used for the generation of chemiluminescence in the presence of a reductant and oxygen It is employed in numerous chemiluminescent assays. Lucigenin is specific for superoxide anion radical.
Formula:
C₂₈H₂₂N₄O₆ MW: 510.51 g/mol |
MDL Number:
MFCD00011925 CAS Number: 2315-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...