Diphenyliodonium nitrate

Supplier: ADIPOGEN CORP MS

CDX-D0046-G050
102987-718EA 296.31 USD
102987-718
Diphenyliodonium nitrate
Diphenyliodonium nitrate

Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.

Formula: C₁₂H₁₀INO₃
MW: 343,12 g/mol
MDL Number: MFCD00031704
CAS Number: 722-56-5
EINECS: 211-962-8
UN: 3087
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR