5(6)-Carboxy-2',7'-dichlorofluorescein
Supplier: ADIPOGEN CORP MS
Synonyms:
Carboxy-DCF
CDX-C0043-G001
CDX-C0043-M200
102987-620EA
600.92
USD
102987-620
102987-618
5(6)-Carboxy-2',7'-dichlorofluorescein
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF)
Fluorescent intracellular pH indicator in the pH 4-5 range.
Formula:
C₂₁H₁₀Cl₂O₇ MW: 445.204 g/mol |
MDL Number:
MFCD00037452 CAS Number: 111843-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...