3-Bromo-1-butyne
Supplier: ADIPOGEN CORP MS
CDX-B0250-G025
CDX-B0250-G005
CDX-B0250-G001
102987-478EA
772.62
USD
102987-478
102987-476
102987-480
3-Bromo-1-butyne
3-Bromo-1-butyne
Building block for synthesis.
Formula:
C₄H₅Br MW: 132.99 g/mol Storage Temperature: 2 - 8°C |
CAS Number:
18668-72-9 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...