Cytochalasin H

Supplier: ADIPOGEN CORP MS

BVT-0447-M001
102989-048EA 131.54 USD
102989-048
Cytochalasin H
Cytochalasin H

Potent mycotoxin. Phytotoxin. Actin polymerization inhibitor. Used in actin polymerization and cytoskeletal reorganization studies. Antibacterial, antifungal, nematocidal and antitumor compound. Apoptosis inducer. Anti-angiogenic agent. Shows immunosuppressive activity. Shown to modulate the CNS with potential anti-parkinson activity.

Formula: C₃₀H₃₉NO₅
CAS Number: 53760-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR