Carboplatin
Supplier: ADIPOGEN CORP MS
Synonyms:
cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II)
Potent platinum-based antineoplastic agent. Analog of cisplatin (Prod. No. AG-CR1-3590) with reduced nephrotoxicity and higher stability. Antitumor agent. Forms inter- and intrastrand DNA adducts/crosslinks, consequently blocking DNA replication and transcription and inducing cell death. Interferes with cell division by mitosis. The damaged DNA elicits DNA repair mechanisms, which in turn activate apoptosis when repair is impossible. Apoptosis inducer. Enhances radiation-induced single-strand DNA breakage.
Formula:
C₆H₁₂N₂O₄Pt MW: 371.26 g/mol |
MDL Number:
MFCD00070464 CAS Number: 41575-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...