cis-Platin

Supplier: ADIPOGEN CORP MS

Synonyms: cis-Diamminedichloroplatinum (II) cis-Diammineplatinum (II) dichloride Platinol

AG-CR1-3590-M250 AG-CR1-3590-M050
102988-682EA 96.92 USD
102988-682 102988-680
cis-Platin
cis-Platin

Potent platinum-based antineoplastic agent. Anticancer compound. Forms inter- and intrastrand DNA adducts/crosslinks, consequently blocking DNA replication and transcription and inducing cell death. Nephrotoxic when used in vivo. Potent chemoimmunotherapeutic. Stimulates the immune responses by activating macrophages and other cells of the immune system. Apoptosis inducer via p53-dependent and -independent mechanisms. Induces apoptosis through caspase-3 activation and XIAP expression inhibition. Targets other proteins and enzymes. Non-competitive NHE-1 inhibitor, an archetypal, membrane-bound mechanosensitive sodium-hydrogen ion transporter.

Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
MDL Number: MFCD00011623
CAS Number: 15663-27-1
UN: 3288
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR