Orion™ Electrode Stand, Thermo Scientific
Supplier: Thermo Scientific
090070
34104-402EA 280.3 USD
34104-402
Orion™ Electrode Stand, Thermo Scientific
This vertical stand with electrode holder can be used as a station for sample containers and vertical adjustment of electrodes.
  • Holds one to three electrodes with a 12mm diameter
  • Vertical motion allows electrodes to be moved up and down into beakers or sample containers
  • Can be used with Orion pH, ORP, and ISE benchtop or portable meters
Recommended for Orion A, A+, Thermo Scientific™ PerpHect™ and pHuture™ Series meters.