    Sort by:
  • Search Within Results
Pseudoanguillosporin A
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Antifungal. Respiration inhibitor. Inhibits mitochondrial respiration in fungi. Binds at the Q0-centre on cytochrome b and blocks the electron transfe...
Small Business Enterprise