You Searched For: Chartreusin
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (102513-442)
Supplier: Adipogen
Description: Antibiotic Antitumor compound. Topoisomerase II inhibitor. Induces single-strand scission in DNA in the presence of reducing agents. Apoptosis inducer...
Small Business Enterprise