    Sort by:
  • Search Within Results
bpV(phen)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Adipogen
Description:   Potent protein phosphotyrosine phosphatase inhibitor. Insulin receptor kinase (IRK) inducer. Insulin mimetic in vitro and in vivo. Apoptosis inducer. ...
Small Business Enterprise