    Sort by:
C-276 alloy
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   High strength, corrosion resistant alloy Ni:Mo:Cr:Fe:W;Co:Mn; 57.5:15.5:15.5:6:3.5:1.5:0.5 approximate wt%