The Vi-CELL™ Cell Counter for Cell Viability Analyzer provides an automatic means to perform the trypan blue dye exclusion method, allowing users to load up to 10 samples at once for easy and automated cell viability analysis. The cellular analysis system can be customized to ensure consistent and accurate cell counts for simple systems like CHO cells and more complex samples like yeast.
- No risk of user bias
- Automation for cell counting, staining, and disposal in minutes
- Easily differentiate live and dead cells for a wide range of cell types
- Programmable cell types with control over the cell sphericity, color, and size
Vi-CELL™ XR software has many cell types preprogrammed, including CHO, Yeast, and Hybridoma
Capacity: 12-position carousel via autoloader
Analysis Range: 2–70 µM
Operating System: Windows 7
It can also design a cellular analysis for a specific cell line.