D-ɑ-Tocopherol succinate

Synonyms: 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acidVitamin E Succinate

V1176-25GM V1176-1KG V1176-5KGBL V1176-500GM
700009-400EA 1300.6 USD
700009-400 700009-402 700009-404 700009-406
D-ɑ-Tocopherol succinate
D-alpha-Tocopherol succinate

Vitamin E Succinate is a potent antioxidant tocopherol shown to enhance the anti-tumor effect of dendritic cell vaccines.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₃H₅₄O₅
MW: 530.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072055
CAS Number: 4345-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR