Victoria blue B

VI105-25GM VI105-100GM
700010-216EA 73.42 USD
700010-216 700010-214
Victoria blue B
Victoria Blue B

Victoria Blue B is used as a nuclear stain in cytology/histochemical applications and can be used in the quantitative determination of phospholipids.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₃H₃₂ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00011878
CAS Number: 2580-56-5

Order Now

Specification Test Results

Absorption Max 599 NM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR