Vetivert oil Indonesian
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
V1222-22.5KG
V1222-500GM
V1222-2.5KG
V1222-100GM
700016-082EA
7313
USD
700016-082
700016-080
700016-084
700016-078
Vetivert oil Indonesian
Vetivert oil
Vetivert Oil, Indonesian is considered a stimulant and tonic, and used in perfumery as well as in cosmetics. The oil, which has an aromatic to harsh woodsy odor, is produced from the roots of a fragrant grass.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
CAS Number:
8016-96-4 |
Specification Test Results
Appearance | Reddish Amber To |
Refractive Index (@ 20°C) | 1.515 - 1.530 |
Specific Gravity (@ 25°C) | 0.983 - 1.022 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...