Triamcinolone diacetate ≥97.0%, micronized USP

Supplier: SPECTRUM CHEMICAL MFG CORP

TR109-10GM TR109-1GM TR109-5GM TR109-100GM TR109-1KGBL
95036-396EA 3924 USD
95036-396 95036-400 95036-398 95036-404 95036-402
Triamcinolone diacetate ≥97.0%, micronized USP
Triamcinolone diacetate

Triamcinolone Diacetate, Micronized, USP is a topical corticosteroid used to treat a variety of skin conditions (e.g., eczema, dermatitis, allergies, rash). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Health hazard

Formula: C₂₅H₃₁FO₈
MW: 478.513 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056462
CAS Number: 67-78-7

Order Now

Specification Test Results

Assay (Anhydrous Basis) 97.0 - 103.0%
Specific Rotation [A]D +39° to +45°
Water Determination max. 6.0%
Residue On Ignition max. 0.5%
Heavy Metals (As Pb) max. 0.0025%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR