trans-1,4-Diaminocyclohexane ≥97%

Synonyms: trans-Cyclohexane-1,4-diaminetrans-Hexahydro-1,4-phenylenediaminetrans-1,4-Cyclohexanediamine

C1940-25GM C1940-100GM
10841-984EA 402.62 USD
10841-984 10841-982
trans-1,4-Diaminocyclohexane ≥97%
trans-1,4-Diaminocyclohexane

Caution: Storage temperature - AIR SENSITIVE, HEAT SENSITIVE: Keep tightly closed. Refrigerate.

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00075174
CAS Number: 2615-25-0
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Purity (GC) 97 %
Purity (Neutralization Titration) 97 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR