Theophylline-7-acetic acid ≥98%

T1299-100GM T1299-25GM T1299-500GM
700009-582EA 161.89 USD
700009-582 700009-584 700009-586
Theophylline-7-acetic acid ≥98%
Theophylline-7-acetic acid

Theophylline-7-acetic Acid is a xanthine derivative.


Non-graded Theophylline-7-acetic Acid supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₁₀N₄O₄
Storage Temperature: Ambient
MDL Number: MFCD00022832
CAS Number: 652-37-9

Order Now

Specification Test Results

Assay 98 % min.
Melting Point 270°-272°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR