Theophyllin, anhydrous ≥97.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
TH110-2.5KG
TH110-100GM
TH110-50KGBL
TH110-500GM
TH110-12KG
95036-254EA
392.6
USD
95036-254
700009-590
95036-252
700009-588
95036-250
Theophyllin, anhydrous ≥97.0% USP
Theophyllin
Theophylline, Anhydrous, USP is an oral medication used to treat asthma and other lung diseases. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Skull and crossbones
UN2811
Formula:
C₇H₈N₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00079619 CAS Number: 58-55-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97.0 - 102.0 % |
Loss On Drying @ 105 C | max. 0.5 % |
Organic Impurities | To Pass Test |
Residue On Ignition | max. 0.15 % |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...