Thiram ≥98%

Synonyms: Tetramethylthiuram disulfideTetramethylthiuram disulphide

T2648-100GM T2648-500GM
10841-978EA 151.9 USD
10841-978 10841-976
Thiram ≥98%
Thiram

Tetramethylthiuram Disulfide, also known as Thiram, is an ectoparasiticide, a fungicide, and an animal repellent used prevent fungal diseases in crops and seeds.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₂N₂S₄
MW: 240.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008325
CAS Number: 137-26-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay Approx. 98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR