Taurine ≥98.5% USP

Synonyms: 2-Aminoethanesulphonic acid2-Aminoethanesulfonic acid

T2860-50KGBL T2860-5KG T2860-100GM T2860-1KG
75812-566EA 1222.55 USD
75812-566 75812-564 75812-562 75812-560
Taurine ≥98.5% USP
Taurine

Taurine, USP is an amino sulfonic acid, but it is often referred to as an amino acid, used in dietary supplements.


All Spectrum Chemical USP dietary supplements are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂H₇NO₃S
MW: 125.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS Number: 107-35-7

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5%
Loss On Drying max.0.3%
Residue On Ignition max. 0.3%
Chloride max. 0.05%
Sulfate max. 0.03%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR