Sulfathiazole ≥99.0%, Powder USP
|
|
Synonyms:
4-Amino-N-(thiazol-2-yl)benzenesulphonamide
, 4-Amino-N-(thiazol-2-yl)benzenesulfonamide
, N1-2-Thiazolylsulphanilamid
, N1-2-Thiazolylsulfanilamid
Sulfathiazole, Powder, USP is a short-acting sulfa drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Exclamation mark
Formula:
C₉H₉N₃O₂S₂ MW: 255.32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005319 CAS Number: 72-14-0 |
Specification Test Results
Assay (Dried Basis) | 99.0 - 100.5% |
Melting Range | 200° - 204°C |
Acidity | To Pass Test |
Loss On Drying | max. 0.5% |
Residue On Ignition | max. 0.1% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...