Sodium sulfanilate dihydrate sodium salt ≥97%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Aminobenzenesulphonic acid, sodium salt dihydrate4-Aminobenzenesulfonic acid, sodium salt dihydrate

S1706-2.5KG S1706-500GM S1706-100GM
700015-774EA 422 USD
700015-774 700015-770 700015-772
Sodium sulfanilate dihydrate sodium salt ≥97%, Reagent Grade
Sodium sulfanilate Dihydrate

Sulfanilic Acid Sodium Salt, Reagent, also known as 4-aminobenzenesulfonic acid, is used to make sulpha drugs and dyes. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₆H₁₀NNaO₅S
MW: 231.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064394
CAS Number: 6106-22-5

Order Now

Specification Test Results

Assay (Corrected For Volatiles) 97 % min.
Loss On Drying @ 105 C 12 - 18 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR