Sodium propionate hydrate ≥99.0% NF
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
S1418-500GM
S1418-45KGBL
S1418-12KG
S1418-2.5KG
700004-748EA
1239
USD
700004-748
95035-914
700004-750
95035-912
Sodium propionate hydrate ≥99.0% NF
Sodium propionate hydrate
Sodium Propionate, NF is a preservative excipient.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
C₃H₅NaO₂·xH₂O MW: 96.06 g/mol (anhy) Storage Temperature: Ambient |
CAS Number:
6700-17-0 |
Specification Test Results
Assay (Dried Basis) | 99.0 - 100.5 % |
Alkalinity | To Pass Test |
Water Determination | max. 1.0 % |
Heavy Metals | max. 10 ppm |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...