SPCMSO194
SO194-2.5KG SO194-45KGBL SO194-12KG SO194-500GM
75812-468EA 1133.83 USD
75812-468 75812-470 95030-886 95030-884
SPCMSO194
Sodium Nitrite, Granular, FCC is used as curing agent and food preservative that creates a distinct flavor, controls lipid oxidation, and acts as an antimicrobial. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Flame over circle;Skull and crossbones;Exclamation mark;Health hazard
UN1500