Sodium nitrate, BiotechGrade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
S1347-500GM
S1347-500GM-CS
75812-254CS
1061.33
USD
75812-254
75812-252
Sodium nitrate, BiotechGrade
Sodium nitrate
Sodium Nitrate, BiotechGrade is a white solid and soluble in water. It is useful in production of glass, fertilizers, pyrotechnics, and food preservatives. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
GHS hazard : Flame over circle;Exclamation mark
UN1498
Formula:
NaNO₃ MW: 84.99 g/mol Density: 2.260 Storage Temperature: Ambient |
MDL Number:
MFCD00085307 CAS Number: 7631-99-4 UN: 1498 ADR: 5.1,III |
Specification Test Results
Appearance | Colorless Granul |
Assay (NaNO₃) | 99.0-100.5 % |
pH of a 1M Solution @ 20°C | 5.5 - 8.0 |
Arsenic (As) | max. 0.0003 % |
Iron | max. 0.0003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...