Sodium metavanadate hydrate, Practical grade

S1338-2.5KG S1338-45KGBL
700001-988EA 2102.88 USD
700001-988 700001-992
Sodium metavanadate hydrate, Practical grade
Sodium metavanadate hydrate

Sodium Metavanadate, Practical is a solid that is yellow in color and soluble in water.


Practical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: NaVO₃·xH₂O
MW: 121.93 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00151340
CAS Number: 20740-98-1
UN: 3285
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR