Sodium iodide ≥99.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Ioduril
, Hydriodic acid sodium salt
, Sodium monoiodide
Sodium Iodide, USP is a crystalline salt that is white and is used as a food supplement to counter and prevent iodine deficiencies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Exclamation mark;Health hazard
Formula:
NaI MW: 149.89 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 UN: 3077 ADR: 9,III |
Specification Test Results
Assay (NaI) | 99.0 - 101.5 % |
Alkalinity | To Pass Test |
Water | max. 2.0 % |
Iodate (IO₃) | To Pass Test |
Nitrate, Nitrite, and Ammonia | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...