Sodium hydrogen fumarate ≥98%

Synonyms: Fumaric Acid Monosodium SaltMonosodium Fumarate

S1313-250GM S1313-50GM
700015-712EA 352.33 USD
700015-712 700015-714
Sodium hydrogen fumarate ≥98%
Sodium hydrogen fumarate

Sodium Hydrogen Fumarate is used as a cosmetic buffering and pH adjusting agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₄O₄
MW: 138.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039097
CAS Number: 5873-57-4

Order Now

Specification Test Results

Assay Approx. 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR