Direct red 80

Synonyms: CI 35780Sirius RedSirius Red F3BPicro-Sirius Red

S1066-25GM S1066-5GM
700010-226EA 120.93 USD
700010-226 700010-224
Direct red 80
Direct Red 80

Sirius Red F3B is a polyazo dye used principally in staining methods for collagen and amyloid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄₅H₂₆N₁₀Na₆O₂₁S₆
MW: 1373.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054389
CAS Number: 2610-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR