Simethicone USP

S1926-100ML S1926-500MLG S1926-4LTGL S1926-500MLG
95035-654EA 128.84 USD
95035-654 700015-808 95035-656 95035-658
Simethicone USP
Simethicone

Simethicone, USP is an antifoaming agent that is used to reduce bloating from excessive gas. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Storage Temperature: Ambient
CAS Number: 8050-81-5

Order Now

Specification Test Results

Assay {[-(CH₃)₂SiO-]n} 90.5 - 99.0 %
Content of Silicon Dioxide 4.0 - 7.0 %
Defoaming Activity To Pass Test
Loss On Heating max. 18.0 %
Heavy Metals max. 5 Ug/G

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR