Sericoside

S1157-1KGBL
700015-612EA 11071.58 USD
700015-612
Sericoside
Sericoside

Sericoside is used in cosmetic antiaging and skin care applications.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 55306-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR