Selenocystamine dihydrochloride, powder

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,2'-Diselanediyldiethanamine dihydrochloride

S2001-100MG S2001-25MG
10841-974EA 127.72 USD
10841-974 10841-972
Selenocystamine dihydrochloride, powder
Selenocystamine dihydrochloride

Caution: Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers

Formula: C₄H₁₄Cl₂N₂Se₂
MW: 318.9934 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035445
CAS Number: 3542-13-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR