Salmon oil Wild Alaskan

Supplier: SPECTRUM CHEMICAL MFG CORP

S1152-2.5KG S1152-200LTB S1152-18KG
700015-598EA 220.93 USD
700015-598 700015-600 700015-596
Salmon oil Wild Alaskan
Salmon oil

Salmon Oil, Wild Alaskan consists primarily of the glycerides of C14-C18 and C16-C22 unsaturated fatty acids and is a cosmetic ingredient for skin and hair conditioning.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 68991-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR