Salmon oil, deodorized Norwegian

Supplier: SPECTRUM CHEMICAL MFG CORP

S1150-200LTB S1150-2.5KG S1150-18KG
700015-590EA 110.42 USD
700015-590 700015-592 700015-588
Salmon oil, deodorized Norwegian
Salmon oil

Salmon Oil, Norwegian, Deodorized consists primarily of the glycerides of C14-C18 and C16-C22 unsaturated fatty acids and is a cosmetic ingredient for skin and hair conditioning.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 68991-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR