Sage oil FCC Spanish Type

S1007-100ML
95030-774EA 351.73 USD
95030-774
Sage oil FCC Spanish Type
Sage oil

SAGE OIL, SPANISH TYPE, FCC, an essential oil, is made from the herb sage and used in culinary applications. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.


GHS hazard : Flame
UN1197

MDL Number: MFCD00217826
CAS Number: 8022-56-8
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Angular Rotation -3 to +24
Refractive Index @ 20°C 1.468-1.473
Saponification Value 14 - 57
Saponification Value After Acetylation 56 - 98
Solubility in Alcohol To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR