Calcium saccharin ≥99.0%, crystals USP

CA167-2.5KG CA167-12KG CA167-125GM CA167-500GM
75810-476EA 552.22 USD
75810-476 75810-918 75810-916 75810-914
Calcium saccharin ≥99.0%, crystals USP
Calcium saccharin

Saccharin Calcium, Crystal, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₄H₈CaN₂O₆S₂·3½H₂O
Storage Temperature: Ambient
CAS Number: 6381-91-5

Order Now

Specification Test Results

Assay (C₁₄H₈CaN₂O₆S₂) 99.0- 101.0 %
Clarity of Solution To Pass Test
Color of Solution To Pass Test
Water Determination max. 15.0 %
Readily Carbonizable Substances To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR