Rosmarinic acid ≥96.5%

Synonyms: (R,E)-3-(3,4-Dihydroxyphenyl)-2-((3-(3,4-dihydroxyphenyl)acryloyl)oxy)propanoic acid

R1062-5GM
700006-590EA 333.85 USD
700006-590
Rosmarinic acid ≥96.5%
Rosmarinic acid

Rosmarinic Acid, a caffeic acid ester, has medicinal and antioxidant applications. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₈H₁₆O₈
MW: 360.318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017740
CAS Number: 20283-92-5

Order Now

Specification Test Results

Assay 96.5 % min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR