Rose water artificial
Supplier: SPECTRUM CHEMICAL MFG CORPSpecification Test Results
Specific Gravity @ 25°C | 0.990 - 1.010 |
Solubility in Water | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...