Renex® 20

R1642-4LT R1642-500ML
700011-074EA 388 USD
700011-074 700011-076
Renex® 20
Renex®

Renex(R) 20 is a low foam surfactant with excellent spot removing properties on hydrophobic fibers.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00132049
CAS Number: 61791-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR