Quinoline yellow SS
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Solvent yellow 33
, 2-(2-Quinolinyl)-1H-indene-1,3(2H)-dione
, Quinoline yellow 2SF
Quinoline Yellow SS, also known as spirit soluble or Solvent Yellow 33, is a yellow dye with a green shade used in polycarbonates and polyamides.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₈H₁₁NO₂ MW: 273.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011593 CAS Number: 8003-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...