Pyrilamine maleate ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Diaminide maleateAnisopyradamineParaminyl maleateMinihist

PY105-5KGBL PY105-100GM
700008-680EA 3737 USD
700008-680 95035-514
Pyrilamine maleate ≥98.0% USP
Pyrilamine maleate

Pyrilamine Maleate, USP is a first generation antihistamine, targeting the H1 receptor, used to treat the common cold. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₂₁H₂₇N₃O₅
MW: 401.463 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069333
CAS Number: 59-33-6

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 100.5 %
Melting Range 99 - 103 °C
Loss On Drying max. 0.5 %
Residue After Ignition max. 0.1 %
Related Compounds To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR