Potassium sorbate ≥98.0%, Powder NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Potassium (2E,4E)-hexa-2,4-dienoate(2E,4E)-2,4-Hexadienoic acid potassium salt2,4-Hexadienoic acid potassium salt(E,E)-2,4-Hexadienoic acid potassium salttrans,trans-2,4-Hexadienoic acid potassium saltPotassium (E,E)-hexa-2,4-dienoateSorbic acid potassium saltPotassium trans,trans-hexa-2,4-dienoate

PO300-45KGBL PO300-12KG PO300-125GM PO300-2.5KG PO300-500GM
89078-974EA 2641.95 USD InStock
89078-974 700005-518 95035-228 95035-230 95035-232
Potassium sorbate ≥98.0%, Powder NF
Potassium sorbate

Potassium Sorbate, Powder, NF is used as an antimicrobial preservative. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: CH₃CH=CHCH=CHCOOK
MW: 150,22 g/mol
Melting Pt: 270 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 590-00-1
EINECS: 246-376-1
Merck Index: 13,07756
https://us.vwr.com/stibo/web/std.lang.all/71/23/2297123.jpg

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 101.0%
Acidity or Alkalinity To Pass Test
Loss On Drying max. 1.0%
Heavy Metals max. 10 ppm
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR